Learn More
Ethanolamine, ACS, 99+%, Thermo Scientific Chemicals
Beschreibung
It is also used in messenger molecules such as palmitoylethanolamide, which have an effect on CB1 receptors. Ethanolamine is used as a scrubber for the removal of methyl bromide. Polyaniline doping ethanolamine is used for modified electrode preparation to measure uric acid in human body with the presence of antiscorbutic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spezifikation
Spezifikation
Melting Point | 10°C |
Density | 1.012 g/mL |
Boiling Point | 169°C to 170°C |
Flash Point | 93°C (199°F) |
Assay Percent Range | >99+% |
Refractive Index | 1.454 |
Quantity | 500 mL |
UN Number | UN2491 |
Beilstein | 505944 |
Sensitivity | Air sensitive; Hygroscopic |
Mehr anzeigen |
Produktvorschläge
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y27K020 | Certificate of Analysis | 02/12/2024 | 036260.AP,036260.AY |
Y27K024 | Certificate of Analysis | 02/12/2024 | 036260.AP,036260.AY |
P19K028 | Certificate of Analysis | 31/03/2024 | 036260.AP,036260.AY |
P19K026 | Certificate of Analysis | 31/03/2024 | 036260.AP,036260.AY |
R13I007 | Certificate of Analysis | 13/10/2022 | 036260.AP,036260.AY |
Sicherheit und Handhabung


- Ethanolamine, ACS
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Ethanolamine
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.