Learn More
Lenz Laborglasintrumente Extractor Heads Acc. to Soxhlet
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05400137
 View more versions of this product
Product Code. 10151521
  • 178.00 € / Each
Estimated Shipment: 26-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Extractor heads acc. to DIN 12602
  • With cone NS 29/32
  • Available with/without NS stopcock
  • Made of DURAN™ tubing
Specifications
Specifications
100 mL
Borosilicate Glass 3.3
NS 45/40
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasintrumente Extractor Heads Acc. to Soxhlet > 100 mL