Learn More
Lenz Laborglasintrumente Extractor Heads Acc. to Soxhlet
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05400131
 View more versions of this product
Product Code. 10681702
  • 164.00 € / Each
Estimated Shipment: 26-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Extractor heads acc. to DIN 12602
  • With cone NS 29/32
  • Available with/without NS stopcock
  • Made of DURAN™ tubing
Specifications
Specifications
70 mL
Borosilicate Glass 3.3
NS 34/35
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasintrumente Extractor Heads Acc. to Soxhlet > 70 mL