missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET RNA Cleanup and Concentration Micro Kit

GeneJET RNA Cleanup and Concentration Micro Kit is designed for rapid and efficient concentration of prepurified RNA samples, as well as for RNA cleanup after DNase I treatment and other enzymatic reactions.

Brand:  Thermo Scientific™ K0841

 View more versions of this product

Product Code. 13203159

  • 233.00 € / 50g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

GeneJET RNA Cleanup and Concentration Micro Kit is designed for rapid and efficient concentration of prepurified RNA samples, as well as for RNA cleanup after DNase I treatment and other enzymatic reactions.

The kit combines the convenience of spin column technology with the selective binding properties of a silica membrane, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions.

Thermo Scientific GeneJET RNA Cleanup and Concentration Micro Kit is designed for rapid and efficient concentration of prepurified RNA samples, as well as for RNA cleanup after DNase I treatment and other enzymatic reactions.

The kit combines the convenience of spin column technology with the selective binding properties of a silica membrane, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions.

Highlights

  • Advanced column design—easy to open, load and handle.
  • Elute in 6 to 10 μL volume.
  • 85 to 100% recovery of a high integrity RNA.
  • Fast and simple—procedure takes less than 5 minutes.

Applications:

  • The purified RNA can be used in common downstream applications
  • RT-PCR, RT-qPCR, Northern blotting and other RNA-based analysis
TRUSTED_SUSTAINABILITY
Specifications

Specifications

For better long-term performance, it is recommended to store the purification columns at 2°C to 8°C.
Spin Column
RNA
Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Nuclease Protection Assays, Nucleic Acid Labeling, Northern Blotting, cDNA Library Construction
GeneJET
Micro
Spin Column
5 min.
Not High-throughput Compatible (Manual)
50 Preps
50 Preps
Total RNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2821616 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberK0841
Lot Number2823945 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK0841
Lot Number2813227 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK0841
Lot Number2805943 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK0841
Lot Number2813226 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK0841
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ GeneJET RNA Cleanup and Concentration Micro Kit > 50 preps

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.