Learn More
Thermo Scientific™ Finntip™ Flex™ Pipette Tips

The Thermo Scientific™ Finntip™ Flex™ Pipette Tips reduce the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI). Ensure a better seal with the pipetter module, for higher accuracy and precision.
69.79 € - 393.00 €
Specifications
Filtered | Non-filtered |
---|---|
For Use With (Equipment) | Thermo Scientific Finnpipette |
Product Line | FinnTip Flex |
Product Code | Brand | Volume (Metric) | Color Code | Fit for Purpose | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Color Code | Fit for Purpose | Price | Quantity & Availability | ||||||
11883420
![]() |
Thermo Scientific™
94060120 |
10 μL | Pink | Certified Quality |
80.70 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
||||||
11863420
![]() |
Thermo Scientific™
94060100 |
10 μL | Pink | Certified Quality |
106.00 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
||||||
11893420
![]() |
Thermo Scientific™
94060310 |
200 μL | Yellow | Certified Quality |
136.00 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
||||||
11873420
![]() |
Thermo Scientific™
94060103 |
10 μL | Pink | Sterile & PCR Ready |
135.00 €
Pack of 960 |
In Stock
Log in to see stock. |
||||||
11803430
![]() |
Thermo Scientific™
94060313 |
200 μL | Yellow | Sterile & PCR Ready |
168.00 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
||||||
11893430
|
Thermo Scientific™
94060316 |
200 μL | Yellow | Non-sterile | N/A | N/A | N/A | |||||
11883430
![]() |
Thermo Scientific™
94060117 |
10 μL | Pink | Certified Quality |
162.61 €
Pack of 1920 |
In Stock
Log in to see stock. ![]() |
||||||
11803440
![]() |
Thermo Scientific™
94060317 |
200 μL | Yellow | Certified Quality |
244.00 €
Pack of 1920 |
In Stock
Log in to see stock. ![]() |
||||||
11813430
![]() |
Thermo Scientific™
94060320 |
200 μL | Yellow | Certified Quality |
69.79 €
Pack of 1000 |
In Stock
Log in to see stock. |
||||||
11738184
![]() |
Thermo Scientific™
94060510 |
300 μL | Orange | Certified Quality |
135.00 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
||||||
11748184
![]() |
Thermo Scientific™
94060513 |
300 μL | Orange | Sterile & PCR Ready |
173.00 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
||||||
11798184
|
Thermo Scientific™
94060516 |
300 μL | Orange | N/A | N/A | N/A | ||||||
11708194
![]() |
Thermo Scientific™
94060517 |
300 μL | Orange | Certified Quality |
177.84 €
Pack of 1920 |
In Stock
Log in to see stock. ![]() |
||||||
11842703
![]() |
Thermo Scientific™
94060520 |
300 μL | Orange | Certified Quality |
80.75 €
Pack of 1000 |
In Stock
Log in to see stock. |
||||||
11458479
![]() |
Thermo Scientific™
94060710 |
1000 μL | Blue | Certified Quality |
177.00 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
||||||
11892703
![]() |
Thermo Scientific™
94060713 |
1000 μL | Blue | Sterile & PCR Ready |
164.00 €
Pack of 960 |
In Stock
Log in to see stock. |
||||||
11865863
|
Thermo Scientific™
94060716 |
1000 μL | Blue | N/A | N/A | N/A | ||||||
11875863
![]() |
Thermo Scientific™
94060717 |
1000 μL | Blue | Certified Quality |
157.00 €
Pack of 1536 |
In Stock
Log in to see stock. ![]() |
||||||
10059682
![]() |
Thermo Scientific™
94060810 |
1200 μL | Turquoise | Certified Quality |
196.00 €
Pack of 960 |
In Stock
Log in to see stock. |
||||||
11742245
![]() |
Thermo Scientific™
94060813 |
1200 μL | Turquoise | Sterile & PCR Ready |
227.25 €
Pack of 960 |
In Stock
Log in to see stock. ![]() |
||||||
11813440
|
Thermo Scientific™
94060816 |
1200 μL | Turquoise | N/A | N/A | N/A | ||||||
11823440
![]() |
Thermo Scientific™
94060817 |
1200 μL | Turquoise | Certified Quality |
393.00 €
Pack of 1536 |
In Stock
Log in to see stock. ![]() |
||||||
10774994
![]() |
Thermo Scientific™
94060820 |
1200 μL | Turquoise | Certified Quality |
101.87 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
||||||
Description
Finntip™ Flex™ Pipette Tips are designed to fit Finnpipette™ F1, F2 and Novus pipettes. They enable you to pipette accurately and precisely, with low liquid retention. The compatibility between the tip and the pipette is indicated with color codes. These Flex™ tips have an ergonomic design that lower the risk of RSI and use less plastic.
Key Features:
- Excellent for single and multichannel pipetting
- Smooth, uniform inner surface of the tips prevents liquid retention
- Available as Sterile or Non-sterile option
- Packaging options: racked, bulk bag or reload tower
- Sterile tips are supplied in vacuum sealed, sterilized, 10 x 96 rack
Softer Finntip Flex Tip
- The softer tip collar reduces the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI)
- It also ensures a better seal with the pipetter module, ensuring higher accuracy and precision
Proven quality
- Sterile options certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Versatile Tip Range
- Available in six volume ranges: 0.2-10 μL, 1-200 μL, 5-300 μL, 100-1000 μL, 50-1200 μL and 10mL Extended (bottle tip)
Immediate traceability
- Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip Flex pipette tip rack insert
- Color-coding visible in rack and reload packaging options
Note:
Availability may vary by country.
Specifications
Non-filtered | |
Thermo Scientific Finnpipette | |
FinnTip Flex |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.