missing translation for 'onlineSavingsMsg'
Learn More
Learn More
First Aid Kit for Sports
Keep a First Aid Kit for Sports on hand to administer first aid to small sports teams and for personal use. The green kit is portable and is accessible for immediate use.
Brand: Safety First K367T/
This item is not returnable.
View return policy
Description
- Dimensions: 250 x 180 x 70mm
- Includes foil blanket, resusciade, instant cold pack, eye wash pods, plasters, dressings, bandages, tape, gloves and guidance leaflet
Specifications
250 mm | |
Green | |
1 Ea. | |
180 mm |
First Aid Kit | |
First Aid Guidance Leaflet, 2 Hypacover first aid dressing assorted, Hypacover Eye Dressings, Hypaband Triangular Bandage, Hypaband conforming Bandage 7.5cm x 4m, Hypaband crepe bandage assorted, Hypaplast microporous tape, 2 Hypatouch disposable gloves p | |
250 x 180 x 70 mm | |
70 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
First Aid Kit for Sports >
Spot an opportunity for improvement?Share a Content Correction