Learn More
Description
Traceable™ Hot Wire Anemometer/Thermometer is ideal for cleanrooms and environmental tests.
- Accuracy is ±1% full scale plus 1 digit
- Displays temperature in user-selectable °C or °F; temperature range from 0°C to 50°C (32°F to 122°F) with a resolution of 0.1°C or °F and accuracy of ±0.8°C (1.5°F)
- Serial RS-232 output allows the meter to be connected to any IBM™-compatible computer or datalogger
- Information can be imported to an existing database and, with the accessory Data Acquisition System, captured, displayed on monitor, and stored†
- Memory function recalls the highest and lowest readings and HOLD button freezes display for later reading
- Measures air velocity in five ranges: 0.2 to 20 mps with a resolution of 0.1 mps
- 0.7 to 72 kph with a resolution of 0.1 kph
- 40 to 3940 fpm with a resolution of 1 fpm
- 0.5 to 44.7 mph with a resolution of 0.1 mph
- 0.4 to 38.8 knots with a resolution of 0.1 knots
†Unit is equipped with a sample program that allows you to store 100 readings; the complete DAS-3™ Data Acquisition System or DAS-4™ Data Logger System is required to transfer these readings to a computer file
Order Info
Dimensions (L x W x H): 7 x 3 x 1.25 in. (17.8 x 7.6 x 3.2 cm); weighs 9.50 oz. (269 g). Available accessories include DAS-4 Data Logger System (06-662-72) and DAS-3 Data Acquisition System (11-661-22).
Specifications
Hot Wire Anemometer/Thermometer | |
Plastic | |
An individually-numbered Traceable™ Certificate is provided with each unit. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Unit is supplied complete with dual thermistors probe, 40-inch cable, Traceable™ Certificate, 6 AAA batteries, and computer output. Size is 7 x 3 x 1-1/4 inches and weight is 9-1/2 ounces. | |
1.25 in. | |
7 in. | |
7.6 cm | |
0°C to 50°C | |
269 g |
±1% | |
Air | |
7 x 3 x 1.25 in. (17.8 x 7.6 x 3.2 cm) | |
3.2 cm | |
17.8 cm | |
32°F to 122°F | |
9.5 oz. | |
3 in. |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.