missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Horizontal Large Gel Systems: Microplate Combs

For large gel systems

Brand:  Fisherbrand™ A2-MT2C

 View more versions of this product

Product Code. 12483657

  • 58.10 € / Each
Estimated Shipment: 01-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Owl A2 and A5 Large Gel Electrophoresis System
Microplate
0.1 cm
0.09 in.
Micro Well Comb
42
13 μL
0.25 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Horizontal Large Gel Systems: Microplate Combs > Comb; 42-tooth; 1.0mm thick; Microplate format; For horizontal large gel systems

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.