missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 96-Well Polypropylene Microplates

Ideal for storage—feature generous 500μL well volumes

Brand:  Fisherbrand™ 235600

72.84 EUR valid until 2025-04-30
Use promo code "24163" to get your promotional price.


 View more versions of this product

Product Code. 11937954

  • 82.35 â‚¬ / Pack of 80
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Polypropylene
  • Minimum binding
  • Resistant to many reagents
  • Withstand temperatures from -80° to +121°C (-112° to +250°F)
  • Automation-friendly design optimized for HTS

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
Blue
96
Round
8 x 12
Polypropylene
Non-sterile
For storage feature generous 500ÎĽL well volumes
Product Suggestions

Product Suggestions

Videos

Videos

  • Fisherbrand 96 Well Polypropylene Microplates
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Fisherbrand 96 Well Polypropylene Microplates

    • Fisherbrand - Advantage aspiration system
      The Fisherbrand aspiration Advantage system is a compact and portable unit with an integrated resistant diaphragm pump, used for the safe and precise aspiration of non-flammable chemical, biological, and medical liquids.
      Video Player is loading.
      Current Time 0:00
      Duration 1:38
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 1:38
       
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected

      Fisherbrand Advantage aspiration system

    SDS
    Documents Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Fisherbrand™ 96-Well Polypropylene Microplates > Round bottom; Non-sterile; Blue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.