missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Samco™ Standard Disposable Transfer Pipettes
GREENER_CHOICE

Single squeeze draws up to 3.2 mL

Brand:  Thermo Scientific™ 225

20.04 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 10100630

  • 25.05 â‚¬ / Pack of 500
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Single squeeze draws up to 3.2 mL

Unbreakable all-in-one pipettes eliminate the hazards of broken glass and exposure to infectious materials.

  • Put an end to matching rubber bulb with glass pipette. Since entire unit is disposable, these pipettes prevent cross-contamination to samples and reduce airborne contamination to personnel
  • One-piece design molded from see-through, low-density polyethylene
  • Inert to biological fluids and most acids
  • Work well anywhere there is a need for quick, safe transfer of fluids
  • In four ready-to-use styles
  • One squeeze draws from 2.0 to 3.3 mL (depending on style used)
  • All deliver repeatable drop sizes
  • Can be sealed and refrigerated for sample storage (seal with Cat. No. 02-678 Hemato-Seal™ Tube Sealant)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

3.2 mL
Research
500/Pk.
15.5 cm
Single-use
Polyethylene
Yes
7.5 mL
Transfer Pipet
Product Suggestions

Product Suggestions

Videos

Videos

  • Thermo Scientific Samco - Graduated transfer pipettes
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Thermo Scientific Samco Graduated transfer pipettes

    • Thermo Scientific Samco - Graduated transfer pipettes
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
       
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected

        Thermo Scientific Samco Graduated transfer pipettes

      SDS
      Documents

      Documents

      Certificates

      Certificates

      A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

      5 results found
      Lot Number Certificate Type Date Catalog Number
      Lot Number1425236 Certificate TypeCertificate of Compliance Date26/03/2025 Catalog Number225
      Lot Number1424041 Certificate TypeCertificate of Compliance Date22/03/2025 Catalog Number225
      Lot Number1412195 Certificate TypeCertificate of Compliance Date22/03/2025 Catalog Number225
      Lot Number1424045 Certificate TypeCertificate of Compliance Date12/03/2025 Catalog Number225
      Lot Number1410958 Certificate TypeCertificate of Compliance Date08/03/2025 Catalog Number225
      Special Offers

      Special Offers

      Product Content Correction

      Your input is important to us. Please complete this form to provide feedback related to the content on this product.

      Product Title
      Thermo Scientific™ Samco™ Standard Disposable Transfer Pipettes > Graduated; Length: 6 in.; Capacity: 7.5 mL

      By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

      Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.