missing translation for 'onlineSavingsMsg'
Learn More

3M™ Doodlebug™ Floor Buffing Pads

For use with the Doodlebug™ Floor Cleaning System

Brand:  3M™ Doodlebug™ 7100194315

Product Code. 17108860

  • 208.00 € / Pack of 30
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Pads are highly durable and easy to clean
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Floor Pads
254 x 118 mm
Doodlebug™ Floor Cleaning System
30 Pk.
Red
Cleaning/Polishing Floors
254 mm
118 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ Doodlebug™ Floor Buffing Pads > Red

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.