Learn More

- Home
- Products
- Buffers and Standards
- Spectroscopy Standards
- AA and ICP Standards
- Gadolinium for AAS 1g/L HNO3 matrix, Fisher chemical™
Gadolinium for AAS 1g/L HNO3 matrix, Fisher chemical™
Product Code. 12983155

Product Code. 12983155
Brand: Fisher Chemical B 021 500
Specifications
Chemical Identifiers
CAS | 7697-37-2 |
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Specifications
Packaging | Bottle |
Quantity | 500 mL |
Physical Form | Liquid |
Concentration | 1 g/L ±0.002 g/L |
Type | Flame Single-element Standard Solution |
Solvent or Matrix | 2% HNO3 |
Chemical Name or Material | Gadolinium |
Product Suggestions
You may be interested in these alternatives
Customers who viewed this item also viewed. This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
-
Catalog No. 12328667
682,00 € / 200 tablets -
Catalog No. 10316380
67,75 € / 2.50 L -
Catalog No. 10666421
65,90 € / 475mL -
Catalog No. 10596814
90,45 € / 50mL -
Catalog No. 10304980
160,00 € / 2.50 L -
Catalog No. 10675492
71,30 € / 1 L -
Catalog No. 10683701
27,55 € / 250mL -
Catalog No. 10640863
197,00 € / Pack of 6 -
Catalog No. 10103740
21,11 € / 100mL -
Catalog No. 10042460
Lead standard solution, for AAS, 1mg/mL Pb in 2% HNO Lead standard solution, for AAS, 1mg/mL Pb in 2% HNO
23,82 € / 100mL -
Catalog No. 10666022
19,10 € / 100mL
Your input is important to us. Please complete this form to provide feedback related to the content on this product.