Learn More
Cytiva GammaBind™ Plus Sepharose™ Medium
Engineered for better binding to rat and mouse IgG subclasses
Brand: Cytiva 17-0886-02
Description
- Recombinant Protein G type 3 coupled to Sepharose CL-6B for easy binding and fast separation
- More enhanced binding capabilities than GammaBind G at both laboratory and process scale
- Yields up to 18mg/mL mouse IgG
- Binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse

Specifications
GammaBind Plus Sepharose | |
90 μm | |
25 mL |
Recombinant GammaBind G type 3 lacking albumin-binding region | |
2°C to 8°C, 20% Ethanol |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- GammaBind Plus Sepharose, 25 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.