Learn More
Thermo Scientific™ GeneJET Genomic DNA Purification Kit
Thermo Scientific GeneJET Genomic DNA Purification Kit is designed for rapid and efficient purification of high quality genomic DNA from various mammalian cell culture and tissue samples, whole blood, bacteria and yeast.
Brand:  Thermo Scientific™ K0721
 View more versions of this product
Product Code. 10304400
  • 197.00 € / Each
Explore more special offers
Description
Description
Thermo Scientific GeneJET Genomic DNA Purification Kit is designed for rapid and efficient purification of high quality genomic DNA from various mammalian cell culture and tissue samples, whole blood, bacteria and yeast.
The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation.
The standard procedure takes less than 20 minutes following cell lysis and yields purified DNA greater than 30 kb in size. Isolated DNA can be used directly in PCR, Southern blotting and enzymatic reactions.
Highlights
Universal—can be used for both cell and tissue samples from a wide range of sources
Efficient—high yields of high molecular weight genomic DNA
Fast—20 minute procedure following cell lysis
Convenient—spin columns are capped and assembled with collection tubes
Applications
Isolated genomic DNA is ideal for use in common molecular biology procedures, including:
• PCR and qPCR
• FastDigest and conventional restriction digestion
• Southern blotting
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Spin Column
Spin Column
Real-Time Quantitative PCR (qPCR), Cloning, Southern Blotting, Sequencing, PCR
GeneJET
50 Preps
<20 min.
Spin Column
Bacteria, Buccal Samples, Cells, Tissue, Yeast
Not High-throughput Compatible (Manual)
Genomic DNA Purification Kit
Genomic DNA
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2817444 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK0721
Lot Number2817518 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK0721
Lot Number2802975 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK0721
Lot Number2809921 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK0721
Lot Number2803024 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK0721
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ GeneJET Genomic DNA Purification Kit > 50 Preps