Learn More
Hirschmann™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation are manufactured from durable DURAN™ glass tubing.
Brand:  Hirschmann™ 2820185
 View more versions of this product
Product Code. 11782142
  • 77.60 € / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With NS ground and polyethylene stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Specifications
DIN EN ISO 1042
200 mL
210 mm
14/23 NS Standard Ground
Yes
Polyethylene
Borosilicate Glass, Duran Glass
±0.15 mL
±0.15 mL
A
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hirschmann™ Class A Volumetric Flasks with Poly Stopper, Blue Graduation > Capacity: 200mL; Socket Joint: 14/23 NS Standard Ground