Learn More
Sartorius Glass Fiber Extraction Thimbles
Extraction thimbles been designed to be compatible with the Soxhlet apparatus, making them ideal for a variety of applications. Designed for versatility, the thimbles can be used with any extractor, including models like the Tectator, to facilitate efficient and high-yield extraction procedures.
208.00 € - 381.00 €
Product Code | Brand | Diameter (Metric) Inside | Length (Metric) | Wall Thickness | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Length (Metric) | Wall Thickness | Price | Quantity & Availability | ||||
30052903
![]() |
Sartorius
FT-1204-019090 |
19 mm | 90 mm | 1.2 mm |
208.00 €
Pack of 25 |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||
30052901
![]() |
Sartorius
FT-1204-022080 |
22 mm | 80 mm | 1.6 mm |
208.00 €
Pack of 25 |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||
30052911
![]() |
Sartorius
FT-1204-025100 |
25 mm | 100 mm | 1.6 mm |
208.00 €
Pack of 25 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||
30052916
![]() |
Sartorius
FT-1204-026060 |
26 mm | 60 mm | 1.5 mm |
208.00 €
Pack of 25 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||
30052906
![]() |
Sartorius
FT-1204-030100 |
30 mm | 100 mm | 1.5 mm |
245.00 €
Pack of 25 |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||
30052905
![]() |
Sartorius
FT-1204-033094 |
33 mm | 94 mm | 1.5 mm |
245.00 €
Pack of 25 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||
30052918
![]() |
Sartorius
FT-1204-043123 |
43 mm | 123 mm | 1.7 mm |
381.00 €
Pack of 25 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||
Description
- Composed of high-purity glass fiber for uncontaminated results
- Engineered for superior mechanical stability to withstand rigorous use
- Glass fiber thimbles offer remarkable resistance to high temperatures
- Designed for a precise fit across a diverse range of extractors
- Exhibits outstanding chemical resistance, ensuring reliability in various extraction processes
Specifications
Glass Fibers | |
G400 | |
Extraction Thimbles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Sartorius Glass Fiber Extraction Thimbles