missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Straw Pipets, Polypropylene, Sterile

Decrease cross contamination risks using disposable pipets
46.22 € - 444.69 €
Product Code | Brand | Volume (Metric) | Length (Metric) | Packaging Type | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Length (Metric) | Packaging Type | Quantity | Price | Quantity & Availability | |||
12602255
![]() |
Corning™
EPI24B-02 |
2.2 mL | 24 cm | 50/Bag | 6000/Cs. |
311.16 €
Pack of 6000 |
Estimated Shipment: 02-09-2025
Log in to see stock. |
|||
11562772
![]() |
Corning™
EPI1B-01 |
1 mL | 19 cm | 40 Bags/25 Units | 1000 |
46.22 €
Pack of 1000 |
In Stock
Log in to see stock. ![]() |
|||
11825380
![]() |
Corning™
EPI24B-01 |
2.2 mL | 24 cm | 25/Bag | 6000/Cs. |
366.15 €
Pack of 6000 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||
12672275
![]() |
Corning™
EPI5-01 |
5 mL | 7.3 cm | 10/Bag | 3600/Cs. |
353.09 €
Pack of 3600 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||
11962785
![]() |
Corning™
EPI1B-05 |
1 mL | 19 cm | 100/Bag | 100/Bag |
444.69 €
Pack of 12000 |
In Stock
Log in to see stock. ![]() |
|||
Description
- Product traceability on each bag
- Sterile 10-3
- Packaged individually in tamper evident single zip bags
Specifications
Polypropylene | |
Straw Pipet | |
Single-use |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ Straw Pipets, Polypropylene, Sterile