missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ Scott™ Luxury Foam Everyday Use Hand Cleanser
Cares for your skin and improves personal hygiene. Kleenex™ Luxury Foam Everyday Use Hand Cleanser keeps costs down and reduces waste.
Brand: Kimberly-Clark™ 6346
63.40 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Ideal for busy washrooms, cleaning hands and maintaining hygiene
- Dispenses the ideal amount of soap for each wash, preventing overuse
- Delivers up to twice as many washes per litre as conventional liquid soaps
- Comes in a strong, hygienically sealed pouch with integrated pump, preventing contamination

Specifications
Hand Cleanser | |
Foam | |
200 mL | |
2.7 kg |
Pink | |
6.8 oz. | |
6 lb. |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Kimberly-Clark™ Scott™ Luxury Foam Everyday Use Hand Cleanser > Pink, Fragrance: Floral, Volume: 200 mL
Spot an opportunity for improvement?Share a Content Correction