missing translation for 'onlineSavingsMsg'
Learn More

TelLab Hand Sanitiser Rub

TelLab Hand Sanitiser Rub is an alcohol-based hand sanitizer formulated according to WHO.

Brand:  TelLab SI-032-1614

Encompass

Product Code. 16650014

  • 41.24 € / 5 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Alert:

Use biocides safely. Always read the label and product information before use.

This item is not returnable. View return policy

Description

Description

  • Ethanol based 78% v/v
  • WHO formulation
  • Packed in stackable 5 L containers
Specifications

Specifications

Hand Sanitizer
Ethanol, glycerol, colorant and perfume free
5 L
WHO formulation, proven efficacy according to EN 1500, Listed in Irish Register of approved PT 1 Human Health Products, ISO 9001 accredited production facility
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
TelLab Hand Sanitiser Rub

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.