missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ HATU Coupling Reagent

This is a coupling reagent for use in peptide synthesis.

Brand:  Applied Biosystems™ GEN076523

 View more versions of this product

Product Code. 15890913

This item is not returnable. View return policy

Description

Description

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in refrigerator (2°C to 8°C).
Peptide Synthesis Resin
Wet Ice
Peptide Synthesis
25 g
Resin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number18G009 Certificate TypeCertificate of Analysis Date02/08/2018 Catalog NumberGEN076523
Lot Number14G013 Certificate TypeCertificate of Analysis Date19/07/2017 Catalog NumberGEN076523
Lot Number12H022 Certificate TypeCertificate of Analysis Date13/04/2017 Catalog NumberGEN076523
Lot Number16J001 Certificate TypeCertificate of Analysis Date13/04/2017 Catalog NumberGEN076523
Lot Number12G023 Certificate TypeCertificate of Analysis Date13/04/2017 Catalog NumberGEN076523
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ HATU Coupling Reagent > 25 g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.