Learn More
Keystone Cleanroom Products Heavy Duty Sterilization Wraps
These wrappers provide an excellent microbial barrier. Ideal for pharmaceutical production applications. For use in steam, ethylene oxide and hydrogen peroxide sterilization processes.
313.00 € - 1180.00 €
Specifications
Product Type | Sterilization Wrap (Heavy-duty) |
---|---|
Basis Weight | 50 gsm |
Autoclavable | Autoclavable |
Certifications/Compliance | CoC |
Closure Type | None |
Product Code | Brand | Length (Metric) | Length (English) | Width (Metric) | Width (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Length (English) | Width (Metric) | Width (English) | Price | Quantity & Availability | |||
17882517
![]() |
Keystone Cleanroom Products
KCRP1-300-0808 |
20.3 cm | 8 in. | 20.3 cm | 8 in. |
371.00 €
Pack of 500 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
17822527
![]() |
Keystone Cleanroom Products
KCRP1-300-0909 |
22.9 cm | 9 in. | 22.9 cm | 9 in. |
394.00 €
Pack of 500 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
17892507
![]() |
Keystone Cleanroom Products
KCRP1-300-0912 |
30.5 cm | 12 in. | 22.9 cm | 9 in. |
478.00 €
Pack of 500 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
17832527
![]() |
Keystone Cleanroom Products
KCRP1-300-1212 |
30.5 cm | 12 in. | 30.5 cm | 12 in. |
470.00 €
Pack of 500 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
17822517
![]() |
Keystone Cleanroom Products
KCRP1-300-1515 |
38.1 cm | 15 in. | 38.1 cm | 15 in. |
585.00 €
Pack of 500 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
17852527
![]() |
Keystone Cleanroom Products
KCRP1-300-1818 |
45.7 cm | 18 in. | 45.7 cm | 18 in. |
805.00 €
Pack of 500 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
17812517
![]() |
Keystone Cleanroom Products
KCRP1-300-2020 |
50.8 cm | 20 in. | 50.8 cm | 20 in. |
928.00 €
Pack of 500 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
17842527
![]() |
Keystone Cleanroom Products
KCRP1-300-2424 |
61 cm | 24 in. | 61 cm | 24 in. |
313.00 €
Pack of 125 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
17832517
![]() |
Keystone Cleanroom Products
KCRP1-300-3030 |
76.2 cm | 30 in. | 76.2 cm | 30 in. |
508.00 €
Pack of 125 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
17862527
![]() |
Keystone Cleanroom Products
KCRP1-300-3636 |
91.4 cm | 36 in. | 91.4 cm | 36 in. |
697.00 €
Pack of 125 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
17842517
![]() |
Keystone Cleanroom Products
KCRP1-300-4040 |
101.6 cm | 40 in. | 101.6 cm | 40 in. |
892.00 €
Pack of 125 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
17882527
![]() |
Keystone Cleanroom Products
KCRP1-300-4545 |
114.3 cm | 45 in. | 114.3 cm | 45 in. |
1060.00 €
Pack of 125 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
17852517
![]() |
Keystone Cleanroom Products
KCRP1-300-4548 |
121.9 cm | 48 in. | 114.3 cm | 45 in. |
1180.00 €
Pack of 125 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
17872527
![]() |
Keystone Cleanroom Products
KCRP1-300-4848 |
121.9 cm | 48 in. | 121.9 cm | 48 in. |
1180.00 €
Pack of 125 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
17872517
![]() |
Keystone Cleanroom Products
KCRP1-300-5454 |
137.2 cm | 54 in. | 137.2 cm | 54 in. |
502.00 €
Pack of 50 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
17892527
![]() |
Keystone Cleanroom Products
KCRP1-300-5472 |
183 cm | 72 in. | 137.2 cm | 54 in. |
779.00 €
Pack of 50 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
17862517
![]() |
Keystone Cleanroom Products
KCRP1-300-5490 |
229 cm | 90 in. | 137.2 cm | 54 in. |
528.00 €
Pack of 25 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
17802537
![]() |
Keystone Cleanroom Products
KCRP1-300-6060 |
152.4 cm | 60 in. | 152.4 cm | 60 in. |
820.00 €
Pack of 50 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
17822537
![]() |
Keystone Cleanroom Products
KCRP1-300-6080 |
203.2 cm | 80 in. | 152.4 cm | 60 in. |
436.00 €
Pack of 25 |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||
Description
- Manufactured and packaged in a certified ISO8 Cleanroom
- 2-Ply
- Clean, no cellulose
- Strong, rip and tear resistant 50 gsm SMS material
- An extensive range of sizes available means less cutting
- Hydrophobic barrier allows sterilization gasses, but prevents liquid and microbial penetration
- Sub packed into sealed labeled poly bags with lot number and date of manufacture for complete traceability
- Certificate of Conformance sent with each shipment
- Made in the USA
sterilization, sterilization sheets, sterilization wraps, autoclavable wraps
Specifications
Sterilization Wrap (Heavy-duty) | |
Autoclavable | |
None | |
Single-use | |
Non-woven Fabric | |
50 gsm | |
50 gsm |
50 gsm | |
CoC | |
Light Blue | |
Steam Sterilization | |
Non-sterile | |
Yes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Keystone Cleanroom Products Heavy Duty Sterilization Wraps