Learn More
Cytiva HiTrap™ Capto™ Q ImpRes IEX Columns
High-throughput medium for intermediate and polishing protein purification
Brand:  Cytiva 17-5470-51
 View more versions of this product
Product Code. 10734572
  • 167.00 € / Pack of 5
Explore more special offers
Description
Description
  • Combined properties of high binding capacity, high flow rate and low back pressure reduces process cycle times and improves productivity at process scale
  • Rigid agarose matrix enables high bed heights and purification of viscous samples at high flow rates
  • Screening of selectivity, binding and elution conditions, as well as small scale purifications
  • Strong quartenary amine anion exchanger
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HiTrap Capto Q ImpRes, 5 x 1 mL
HiTrap Capto Q ImpRes chromatography column is packed with a strong anion exchange modern resin for high resolution, small-scale protein purification.
7 mm
5 x 1 mL
<4 mL/min.
4°C to 30°C in 20% Ethanol
1 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva HiTrap™ Capto™ Q ImpRes IEX Columns > HiTrap Capto Q ImpRes prepacked column; Pack Size: 5 x 1mL