Learn More
Gibco™ Human FGF-7 (KGF) Recombinant Protein, PeproTech®
Recombinant Protein
Brand:  Gibco™ 100-19-250UG
1750.26 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
 View more versions of this product
Product Code. 17850783
  • 1882.00 € / 250µg
Explore more special offers
Description
Description
Recombinant Human KGF (FGF-7) is an 18.9 kDa protein consisting of 163 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Gibco™ Human FGF-7 (KGF) Recombinant Protein, PeproTech® > 250 μg; Unlabeled