Learn More
Invitrogen™ Human ZMYM1 Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP1203
Product Code. 15205209
  • 141.00 € / 50µg
Explore more special offers
Description
Description
ZMYM1 Synthetic Peptide, PEP-1203, from Invitrogen.
Zinc-finger proteins contain DNA-binding domains characterized by the unique role of zinc and have a wide variety of functions such as transcriptional activation or repression. The protein folding and the DNA binding ability are governed by the coordination of a zinc ion. As a member of the MYM (myeloproliferative and mental retardation) gene family, ZMYM1 is widely expressed in different tissues in eukaryotes under several forms derived by alternative splicing. While its function remains unknown, the related protein ZMYM2 has been shown to associate with and stabilize the LSD1-CoREST-HDAC1 (LCH) complex of chromatin through its MYM-type zinc fingers, thereby enhancing the transcriptional repression of several genes, suggesting that ZMYM1 may play a similar role.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
79830
purified
Human ZMYM1
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q5SVZ6
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
ZMYM1
0.02% sodium azide
50 μg
PBS
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ Human ZMYM1 Synthetic Peptide > 50 μg; Unlabeled