Learn More
Invitrogen™ iBind™ Flex Mini Insert Replacement
Use with the iBind™ Flex Western Device (SLF2000). Invitrogen™ iBind™ Flex Mini Well Insert Replacement is a well insert that customizes the format to process two mini blots. It features two wells on row 1 and 3.
Brand:  Invitrogen™ SLF2002
Product Code. 15401254
  • 279.00 € / 1 set
Explore more special offers
Description
Description
The iBind™ Flex Western System offers automated western blot processing. Using customized well inserts, it can accommodate various blot formats including midi, mini, and vertically cut strips. Based on Sequential Lateral Flow (SLF) technology, all blocking, antibody incubation and washing steps during immunodetection are hands-free. Compared to manual methods, the iBind Flex Western System requires less primary antibody, enables reproducible results and is compatible with downstream detection protocols.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
iBind
Replacement Insert
1 insert
iBind
Two wells on row 1 and 3
Room Temperature
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ iBind™ Flex Mini Insert Replacement > Mini well insert; With two wells on row 1 and 3