Learn More
Cytiva Nucleon™ HT Genomic DNA Extraction Kit
Phenol-free extraction of genomic DNA from paraffin-embedded sections and hard tissue requiring proteinase K digestion. Consistent extraction of high-quality, amplifiable DNA. Protocols developed for those tissues which don't homogenize easily in lysis buffer and require digestion with proteinase K.
Brand: Cytiva RPN8509
Includes: Reagent B, 18mL; 5M Sodium perchlorate, 6mL; Nucleon resin, 8mL; Proteinase K, 10mg
Description
- Extract DNA from FFPE samples, mouse tail, Xiphisternum etc.
- User-friendly protocol enables efficient processing of large numbers of samples
- Patented Nucleon resin delivers highly purity by binding macromolecules other than DNA
- DNA is not bound to any solid surface throughout the purification avoiding exposure to shearing forces promoting recovery of high yields of highly intact DNA
- Each kit is designed to recover high-quality DNA from 50 preparations of up to 25mg of hard tissue or 50 paraffin sections

Specifications
illustra Nucleon HT enables DNA extraction from hard tissue and paraffin sections. | |
Extract genomic DNA from Paraffin embedded sections and hard tissue requiring Proteinase K digestion | |
illustra Nucleon HT |
Paraffin-embedded Tissue, Difficult Samples | |
18mL Reagent B, 6mL 5M Sodium Perchlorate, 8mL Nucleon™ Resin, 10mg Proteinase K | |
1 Kit |
Safety and Handling
- Danger
Hazard Category
- Oxidising liquid Category 2
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501-Dispose of contents/container to .
Your input is important to us. Please complete this form to provide feedback related to the content on this product.