Learn More
Thermo Scientific™ Immuno Breakable Modules Clear
Choose from four surfaces and a variety of wells and configurations with these immuno breakable modules, which offer an optimal choice of products for the specific assay.
Brand: Thermo Scientific™ 446442
392.74 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Description
Choose from four surfaces and a variety of wells and configurations with Thermo Scientific™ Immuno Breakable Modules, which offer reliability, well-to-well reproducibility and an optimal choice of products for the specific assay.
Choose between LockWell, BreakApart and Breakable 1x8 modules in standard 96-well frames.
Features:
- For solid-phase immunoassays, e.g. ELISA
- Choice of four surfaces for optimal binding
- Free-standing modules are easy to handle
- Alphanumeric system for easy orientation
- Fits standard equipment
- High optical quality for reliable results
- Polystyrene
Choice of Surface:
- PolySorp™, Immulon™ 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
- Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
- MaxiSorp™, Immulon 4 HBX, and Enhanced Binding (EB) are hydrophilic and ideal for antibody sandwich assays
- MultiSorp™ are very hydrophilic and work well for glycans and water-soluble proteins

Specifications
Breakable Strips (LockWell) | |
No | |
Without Lid | |
Colorimetric | |
Inner packs of 5 | |
Strip Plate | |
350 μL | |
250 μL | |
PolySorp | |
For adsorption of hydrophobic molecules | |
5 | |
60/Cs. | |
C8 |
Hydrophobic | |
Strip Plate | |
High optical quality for reliable results | |
8 x 12 | |
Clear | |
Polystyrene | |
96 | |
Non-sterile | |
Round, C Bottom | |
60 | |
None | |
Lipids |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
185534 | Certificate of Compliance | 11/03/2025 | 446442 |
184769 | Certificate of Compliance | 16/01/2025 | 446442 |
165828 | Certificate of Compliance | 03/12/2024 | 446442 |
165747 | Certificate of Compliance | 03/12/2024 | 446442 |
164321 | Certificate of Compliance | 03/12/2024 | 446442 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.