missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NuPAGE™ LDS Sample Buffer (4X)

Used to prepare protein samples for denaturing gel electrophoresis with NuPAGE™ Novex™ gels. Invitrogen™ Novex™ NuPAGE™ LDS Sample Buffer (4X) contains lithium dodecyl sulfate, pH 8.4, which allows for maximum activity of the reducing agent.

Brand:  Invitrogen™ NP0007

16.72 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 11549166

  • 27.41 € / 10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: NuPAGE LDS Sample Buffer (4X), 10mL
This item is not returnable. View return policy

Description

Description

Optimal protein sample preparation for polyacrylamide gel electrophoresis (PAGE) requires denaturing and reducing protein disulfide bonds.

  • Highly viscous and concentrated solution that contains higher a concentration of glycerol
  • Contains twice the amount of LDS compared to the amount of SDS in Novex™ Tris-Glycine SDS Sample Buffer or in Tricine SDS Sample Buffer
  • NuPAGE™ LDS Sample Buffer should be brought to room temperature (25°C) prior to use

Order Info

Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sample Buffer (4X) containing lithium dodecyl sulfate (LDS) at pH 8.5 with SERVA Blue G250 and phenol red.

Store at 4°C to 25°C.

4 X
LDS Sample Buffer
NuPAGE™
Approved for shipment at Room Temperature or on Wet Ice
Sample Loading Buffers
NuPAGE
NuPAGE™ Gels
10mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3016242 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberNP0007_LITL18 PROMO,NP0007
Lot Number3051986 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberNP0007_LITL18 PROMO,NP0007
Lot Number3088892 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberNP0007_LITL18 PROMO,NP0007
Lot Number2463558 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberNP0007_LITL18 PROMO,NP0007
Lot Number2463560 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberNP0007_LITL18 PROMO,NP0007
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ NuPAGE™ LDS Sample Buffer (4X) > 10mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.