Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class F1
Calibrate masses of class II and III weighing instruments. Fisherbrand™ Class F1 Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F2 weights and for use with weighing instruments of special accuracy.
48.00 € - 1050.00 €
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | ||||||
15565049
|
Fisherbrand™
FB67767 |
1 g |
48.00 €
1 unit |
Estimated Shipment: 27-05-2025
Log in to see stock. |
||||||
15535059
|
Fisherbrand™
FB67774 |
200 g |
104.00 €
1 unit |
Estimated Shipment: 27-05-2025
Log in to see stock. |
||||||
15545059
|
Fisherbrand™
FB67775 |
500 g |
198.00 €
1 unit |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
15595049
|
Fisherbrand™
FB67770 |
10 g |
57.00 €
1 unit |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
15575049
|
Fisherbrand™
FB67768 |
2 g |
49.00 €
1 unit |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
15575059
|
Fisherbrand™
FB67778 |
5 kg |
590.00 €
1 unit |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
15585049
|
Fisherbrand™
FB67769 |
5 g |
50.00 €
1 unit |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
15545189
|
Fisherbrand™
FB67905 |
10 kg |
1050.00 €
1 unit |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
15555059
|
Fisherbrand™
FB67776 |
1 kg |
254.00 €
1 unit |
In Stock
Log in to see stock. ![]() |
||||||
15565059
|
Fisherbrand™
FB67777 |
2 kg |
372.00 €
1 unit |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
15525059
|
Fisherbrand™
FB67773 |
100 g |
85.00 €
1 unit |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
15515059
|
Fisherbrand™
FB67772 |
50 g |
68.00 €
1 unit |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
15505059
|
Fisherbrand™
FB67771 |
20 g |
62.00 €
1 unit |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
Description
- Class F1 stainless steel calibration weight
- In compliance with OIML R111
Specifications
F1 | |
Stainless Steel | |
Cylindrical |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class F1