Learn More
Mettler Toledo™ InLab™ Sensor Cables: S7 Connection
Electrode cables to connect Mettler Toledo InLab sensors to any Mettler Toledo meter.
Brand:  Mettler Toledo™ 30281920
98.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15519661
  • 101.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Detachable
  • Resuable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
S7-DIN 19262
To connect Mettler Toledo InLab sensors to any meter
3.0 m
Mettler Toledo InLab Sensors
9.8 ft.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Mettler Toledo™ InLab™ Sensor Cables: S7 Connection > Connector: DIN 19262; Length: 3m (9.8 ft.)