Learn More
Fisherbrand™ Insert for Midi Boxes
Manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes; these serve as an ideal solution for short-term storage of radioactive waste or radioisotopes.
Brand: Fisherbrand™ CSR-R2
Description
- Beta – 32 x 2ml Cryotubes
- Inserts for midi boxes
- Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
- Also supplied is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids
- These serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
- Both the Beta and Gamma storage bins are available in five sizes, while the two largest bin models have wheels for easy transportation
- Optional heavy duty drawstring bags may also be purchased
Specifications
For Midi Boxes | |
Insert | |
160mm |
40mm | |
32 x 2mL |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.