missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Insulin, human recombinant, zinc solution
4 mg/mL
Brand: Gibco™ 12585014
This item is not returnable.
View return policy
Description
Potency: (dry basis) ≥27 USP units/mg.
Suggested range for supplementation: 2 to 20μg/mL.

Specifications
Store in freezer (-5 to -30°C). | |
Mammalian Cell Culture | |
Cell Culture Supplement | |
Dry Ice |
Mammalian Cells | |
Liquid | |
5 mL |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2679035 | Certificate of Analysis | 29/01/2025 | 12585014 |
2753797 | Certificate of Analysis | 29/01/2025 | 12585014 |
2753799 | Certificate of Analysis | 28/01/2025 | 12585014 |
2753801 | Certificate of Analysis | 27/01/2025 | 12585014 |
2861512 | Certificate of Analysis | 27/01/2025 | 12585014 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Insulin, human recombinant, zinc solution > 5mL
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction