Learn More
Invitrogen™ Cellfectin™ II Reagent
Cationic-lipid formulation designed for optimal transfection of insect cells
Brand: Invitrogen™ 10362100
1039.30 EUR valid until 2025-12-31
Use promo code "24270" to get your promotional price.
Includes: Contains one tube (1mL) Cellfectin II Reagent.
Description
Cellfectin II is an improved version of the original Cellfectin Reagent. In addition to providing the same great performance as Cellfectin, Cellfectin II Reagent is manufactured to ensure lot-to-lot consistency, and is optimized to be used in a faster protocol. Transfection with Cellfectin II Reagent leads to consistent and efficient transfection of Sf9, Sf21, and High Five™ cells when using BaculoDirect™ and InsectSelect™ Expression Systems. Cellfectin II Reagent can also be used to transfect adherent or suspension mammalian cells in serum-containing or serum-free media.
PCR & Real-Time PCR, Reverse Transcription
Order Info
Shipping Condition: Wet Ice

Specifications
Contains one tube (1 ml) Cellfectin™ II Reagent. Store at 4°C. Do not freeze. | |
Baculovirus DNA, Plasmid DNA | |
BaculoDirect and InsectSelect Expression Systems | |
Not High-throughput Compatible (Manual) | |
Reagent | |
No |
Insect Cells | |
Lipid-based Transfection | |
Protein Expression and Transfection | |
Cellfectin™ | |
1 mL | |
Wet Ice |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2971218 | Certificate of Analysis | 31/01/2025 | 10362100 |
3035196 | Certificate of Analysis | 31/01/2025 | 10362100 |
2447449 | Certificate of Analysis | 29/01/2025 | 10362100 |
CN2507731 | Certificate of Analysis | 29/01/2025 | 10362100 |
2464585 | Certificate of Analysis | 29/01/2025 | 10362100 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.