missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TrackIt™ 1 Kb Plus DNA Ladder
GREENER_CHOICE

Suitable for sizing linear double-stranded DNA fragments from 100 bp to 12 kb

Brand:  Invitrogen™ 10488085

200.45 EUR valid until 2025-12-31
Use promo code "24358" to get your promotional price.


Product Code. 10540703

  • 230.00 € / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Includes: TrackIt™ 1 Kb Plus DNA Ladder (0.1μg/μL) in 10mM Tris-HCl (pH 7.5), 10mM EDTA (pH 8.0), 0.06% XCFF, 0.6% tartrazine, and 5% glycerol; For 100 applications
This item is not returnable. View return policy

Description

Description

TrackIt™ 1Kb Plus DNA Ladder is prepared from plasmid containing repeats of 1,000 bp DNA fragment. The 1,650bp band is generated from pUC and all bands <1,000 bp are from lambda DNA. TrackIt 1Kb Plus DNA Ladder is formulated with two unique tracking dyes, Xylene Cyanol FF (XCFF) and tartrazine, that allow to visually track DNA migration during electrophoresis and also indicate when maximum resolution is achieved. Tracking dyes do not obscure visualization of DNA bands in ladder as dyes run outside limits of most DNA bands in ladder.

  • Consists of 12 bands in 1,000bp increments ranging in size from 1 to 12 Kb, a 1,650 bp fragment, and vector DNA fragments from 100 to 850 bp
  • Orientation doublet at 1,650 bp and 2,000 bp for easy reference
  • Formulated with unique tracking dyes, XCFF and tartrazine
  • Designed for use with E-Gel™ agarose gels and TBE/TAE agarose gels
  • Supplied in ready-to-load format, stable at room temperature
  • Visualized with ethidium bromide or SYBR™ Green staining
  • Compatibile with agarose gels E-Gel™ gels

Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Approved for shipment at Room Temperature or on Wet or Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 500 µL TrackIt 1 Kb Plus DNA Ladder
• 1 mL 6X TrackIt Cyan/Orange Loading Buffer

Store at room temperature or at 4°C for up to 6 months. For longer periods, store at -20°C.

Agarose
Yes
100 to 15000 bp
Agarose gel
100 Applications
50 μg
Individual chromatography-purified DNA fragments
0.1 μg/μL
DNA Ladder
1 mL
500 μL
Sustainable packaging
TrackIt™
Approved for shipment at Room Temperature or on Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2816445 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10488085
Lot Number2816366 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10488085
Lot Number2794741 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10488085
Lot Number2842109 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10488085
Lot Number2842201 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number10488085
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ TrackIt™ 1 Kb Plus DNA Ladder > 100 applications

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.