Learn More
Invitrogen™ E-Gel™ 1 Kb Plus DNA Ladder

Powerful tool for estimating molecular weight of linear, double-stranded DNA fragments
Brand: Invitrogen™ 10488090
261.25 EUR valid until 2025-12-31
Use promo code "24358" to get your promotional price.
Includes: E-Gel™ 1 Kb Plus DNA Ladder is supplied at 0.035μg/μL in 10mM Tris-HCl (pH 7.5), 1mM EDTA (pH 8.0), 0.0075% XCFF, 0.05% tartrazine, and 5% glycerol. Includes 500μL of the E-Gel 1 Kb Plus DNA Ladder, enough for 100 loads
Description
The E-Gel™ 1Kb Plus DNA ladder is composed of 20 highly purified, double-stranded DNA bands spanning 100 base pairs to 12,000 base pairs. Ladder has 12 evenly spaced bands ranging from 1Kb to 12Kb, quick orientation band at 1,650 base pairs that forms distinct doublet with 2Kb band, and seven bands of round sizes below 1Kb. The E-Gel 1Kb Plus DNA Ladder is specifically formulated with minimal salts to perform exceptionally well on E-Gel™ EX pre-cast agarose gels. Unlike some other DNA ladders, E-Gel 1Kb Plus DNA Ladder has been specially designed to have bands of round, easy-to-remember sizes. Logical band layout, and quick identification doublet, allow quick and easy estimates of band size at glance.
- Round, easy-to-remember band sizes
- Broad size range (100bp–12Kb)
- Formulated specifically for E-Gel EX gels
- Room temperature stable for easy storage
- Single ladder fits most needs
- Ready-to-use formulation for E-Gel and E-Gel EX gels
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Approved for shipment at Room Temperature or on Wet or Dry Ice

Specifications
• 2 x 1 mL E-Gel 1 Kb Plus DNA Ladder • 1 mL 1X E-Gel Sample Loading Buffer Store at room temperature or at 4°C for up to 6 months. For longer periods, store at -20°C. |
|
Agarose | |
Yes | |
100 to 15000 bp | |
E-Gel™ agarose gels | |
Linear DNA | |
Approved for shipment at Room Temperature or on Dry Ice |
0.025 μg/μL | |
DNA Ladder | |
1 mL | |
2 x 1000 μL | |
Sustainable packaging | |
100 applications | |
Individual chromatography-purified DNA fragments |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2863809 | Certificate of Analysis | 31/01/2025 | 10488090 |
3079471 | Certificate of Analysis | 31/01/2025 | 10488090 |
2817813 | Certificate of Analysis | 29/01/2025 | 10488090 |
2817971 | Certificate of Analysis | 29/01/2025 | 10488090 |
2804871 | Certificate of Analysis | 29/01/2025 | 10488090 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.