missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodoacetonitrile, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Iodoacetonitrile is used for alkylation of nucleophiles. It undergoes Reformatsky and Wittig-type reactions with aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Iodoacetonitrile is used for alkylation of nucleophiles. It undergoes Reformatsky and Wittig-type reactions with aldehydes.
Solubility
Soluble in benzene, alcohol, acetone, ether. Slightly soluble in water. Insoluble in hexane.
Notes
Light Sensitive. Store at 4°C. Incompatible with reducing agents, light and bases. Store away from strong bases.
Iodoacetonitrile is used for alkylation of nucleophiles. It undergoes Reformatsky and Wittig-type reactions with aldehydes.
Solubility
Soluble in benzene, alcohol, acetone, ether. Slightly soluble in water. Insoluble in hexane.
Notes
Light Sensitive. Store at 4°C. Incompatible with reducing agents, light and bases. Store away from strong bases.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 2.306 |
Boiling Point | 70°C to 72°C (10 mmHg) |
Flash Point | 86°C (186°F) |
Linear Formula | ICH2CN |
Refractive Index | 1.574 |
Quantity | 50 g |
UN Number | UN2922 |
Beilstein | 1734628 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in benzene,alcohol,acetone,ether. Slightly soluble in water. Insoluble in hexane. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'