missing translation for 'onlineSavingsMsg'
Learn More

Isoamyl nitrite, 96%, stabilized, Thermo Scientific™

Product Code. 10445081
Click to view available options
100g; Plastic bottle packed in tin can
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 110-46-3, 497-19-8
Density 0.8700g/mL
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Plastic Bottle in Tin Can
Molecular Formula C5H11NO2
Linear Formula (CH3)2CHCH2CH2ONO
Refractive Index 1.3840 to 1.3880 (20°C, 589nm)
MDL Number MFCD00002057
Quantity 100g
Beilstein 01, 402
Fieser 01,40; 04,270; 05,358; 06,307; 09,258
Merck Index 15, 5168
Synonym isoamyl nitrite, isopentyl nitrite, amilnitrite, vaporole, aspiral, nitrous acid, 3-methylbutyl ester, 3-methylbutanol nitrite, pentanoli nitris, nitrous acid, isopentyl ester, amyl nitrite i
InChI Key OWFXIOWLTKNBAP-UHFFFAOYSA-N
SMILES CC(C)CCON=O
IUPAC Name 3-methylbutyl nitrite
Stabilizer 0.5% sodium carbonate
Molecular Weight (g/mol) 117.15
PubChem CID 8053
ChEBI CHEBI:2691
Formula Weight 117.15
Percent Purity ≥95%
Chemical Name or Material Isoamyl nitrite, 90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.