Learn More
DuPont™ Tyvek™ IsoClean™ Unhooded Coverall with Bound Neck

Clean-processed to maximize cleanliness and pre-sterilized by gamma irradiation. DuPont™ Tyvek™ IsoClean™ Unhooded Coverall with Bound Neck provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash. Breathable and low linting.
395.00 € - 469.00 €
Specifications
Product Type | Cleanroom Coveralls |
---|---|
Material | HDPE |
Color | White |
Disposable | Single-use |
Waist Style | Elastic at Back |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15694894
![]() |
DuPont™
D15429648 |
Small |
395.00 €
Pack of 25 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||||
15604904
![]() |
DuPont™
D15429659 |
Medium |
395.00 €
Pack of 25 |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
15614904
![]() |
DuPont™
D15429666 |
Large |
395.00 €
Pack of 25 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||||
15624904
![]() |
DuPont™
D15429671 |
X-Large |
395.00 €
Pack of 25 |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
15634904
![]() |
DuPont™
D15429681 |
2X-Large |
395.00 €
Pack of 25 |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
15644904
![]() |
DuPont™
D15429690 |
3X-Large |
469.00 €
Pack of 25 |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
Description
- Available in sizes S to XXXL
- Clean-processed and gamma-sterilized
- Bound internal seams. Elasticated waist at back for good fit
- Tyvek™ covered elasticated thumb loops
- Tunneled elastication at wrists and ankles
- Front zipper closure
- Storm flap
Specifications
Cleanroom Coveralls | |
White | |
Elastic at Back | |
Elastic | |
Bound | |
ISO 11137-1, ISO Class 5 | |
Biotech, Pharmaceutical, Medical Device Manufacturing, Food Processing, Cosmetics Industry. and Other Critical Environments |
HDPE | |
Single-use | |
Elastic | |
Front Zipper | |
Sterile | |
GMP class A/B, Category III, Type 5-B and 6-B, EN 14126, EN 1073-2 | |
25/Pk. |
The user should make the risk assessment to determine the protection required.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.