Learn More
Jenway™ Genova Nano Micro-volume Spectrophotometer
Measures small sample volumes as low as 0.5μL with a high degree of accuracy, reproducibility and speed. Jenway™ Genova Nano Micro-volume Spectrophotometer is ideal for DNA, RNA and protein measurements and can detect DNA concentrations as low as 2ng/μL.
Brand: Jenway™ 83070-02
6348.68 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
The Genova Nano measures small sample volumes as low as 0.5μL with a high degree of accuracy, reproducibility and speed. It is ideal for DNA, RNA and protein measurements and can detect DNA concentrations as low as 2ng/μL. This spectrophotometer has the ability to measure small sample volumes which conserves precious samples, reduces the need for dilutions and eliminates the requirement for cuvettes. Cleaning is quick and simple; wiping the read heads with a microfibre cloth removes all trace of the sample, allowing faster changeover between samples and therefore increasing sample throughput.- Pre-programmed for DNA/RNA analysis
- DNA purity scan across entire wavelength range
- Pre-programmed protein methods
- OD 600 measurement mode
- Only 0.5μL sample volume required
- Press-to-read xenon lamp
- Easy and quick to clean
- Fast measurement time
- Conserves precious samples
- Three year warranty, including xenon lamp
- Standard spectrophotometer functions
- Icon driven software
- Small footprint
- Save methods and results to USB memory stick
Specifications
±2% at 260nm | |
400 mm | |
Microvolume | |
Spectrophotometer | |
220 mm | |
3 Years | |
7.7 kg |
15 to 125 A (10 mm equivalent) | |
Xenon | |
5 nm | |
198 to 1000 nm | |
<6.5sec. | |
±2 nm | |
275 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.