missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L(+)-allo-Threonine, 99% Product Code.: 10041701

Thermo Scientific Chemicals L(+)-allo-Threonine, 99%

Product Code. 10041701
100 mg, Glass Bottle
Click to view available options
Quantity:
100 mg
250 mg
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10041701

Brand: Thermo Scientific Chemicals 198541000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28954-12-3
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064268
InChI Key AYFVYJQAPQTCCC-HRFVKAFMSA-N
Synonym l-allo-threonine, l-allothreonine, 2s,3s-2-amino-3-hydroxybutanoic acid, l +-allo-threonine, allo-l-threonine, h-allo-thr-oh, allothreonine, allo-threonine, unii-hcq253ckvk, dl-allo-threonineShow MoreShow Less
PubChem CID 99289
ChEBI CHEBI:28718
IUPAC Name (2S,3S)-2-amino-3-hydroxybutanoic acid
SMILES C[C@H](O)[C@H](N)C(O)=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula CH3CH(OH)CH(NH2)CO2H
Beilstein 04, III, 1629
Specific Rotation +8° to +10° (20°C, 589nm) (c=2, water)
Specific Rotation Condition + 9.00 (20.00°C c=2,H2O)
Quantity 100 mg
Formula Weight 119.12
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material L(+)-allo-Threonine, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466894 Certificate TypeCertificate of Analysis Date29/10/2024 Catalog Number198541000,198542500
Lot NumberA0398344 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number198541000,198542500
Lot NumberA0270775 Certificate TypeCertificate of Analysis Date07/12/2022 Catalog Number198541000,198542500
Product Identifier
  • L(+)-allo-Threonine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L(+)-allo-Threonine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.