missing translation for 'onlineSavingsMsg'
Learn More
L-leucine, 99.42%, MP Biomedicals™ Product Code.: 11329019

L-leucine, 99.42%, MP Biomedicals™

Product Code. 11329019 Shop All MP Biomedicals Products
5 kg
Click to view available options
Quantity:
5 kg
This item is not returnable. View return policy

Product Code. 11329019

Brand: MP Biomedicals 0210215805

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 61-90-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00002617
InChI Key ROHFNLRQFUQHCH-MLHKIVSYNA-N
Synonym l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acidShow MoreShow Less
PubChem CID 6106
ChEBI CHEBI:15603
SMILES CC(C)C[C@H](N)C(O)=O

Specifications

Color White
Assay Percent Range 0.9942
Density 1.293g/mL
IUPAC Name 2-amino-4-methylpentanoic acid
Quantity 5 kg
Formula Weight 131.18
Physical Form Small Lustrous Plates or Crystalline Powder
Chemical Name or Material L-Leucine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-leucine, 99.42%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.