missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Leucine, 99% Product Code.: 11975331

Thermo Scientific Chemicals L-Leucine, 99%

Product Code. 11975331
2.5 kg
Click to view available options
Quantity:
2.5 kg
This item is not returnable. View return policy

Product Code. 11975331

Brand: Thermo Scientific Chemicals 125120025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 286.0°C to 288.0°C
Ammonium 200ppm max.
CAS 61-90-5
pH 5.5 to 7.0 (1% aq. soln.)
Color White
Chloride 500ppm max.
Heavy Metals (as Pb) 20ppm max.
Infrared Spectrum Authentic
Iron (Fe) 30ppm max.
Assay Percent Range 99%
Loss on Drying 0.2 % max. (105°C, 3 hrs)
Other Amino Acids 0.5 % max. (TLC)
Molecular Formula C6H13NO2
Packaging Plastic Drum
MDL Number MFCD00002617
Specific Rotation + 15.40
Sulfate 300ppm max.
Transmittance 95 % min. (c=10, 2 N HCl, 430nm) 1 cm cell
Sulfated Ash 0.4 % max.
Synonym l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
Solubility Information Solubility in water: 22.4g/L (20°C). Other solubilities: 10.9g/L acetic acid, slightly soluble in alcohol, insoluble in ether
Sublimation Point 145.0 °C
InChI Key ROHFNLRQFUQHCH-MLHKIVSYNA-N
SMILES CC(C)C[C@H](N)C(O)=O
IUPAC Name (2S)-2-amino-4-methylpentanoic acid
Specific Rotation Condition + 15.40 (20.00°C c=4,6N HCl)
Molecular Weight (g/mol) 131.18
Quantity 2.5 kg
PubChem CID 6106
ChEBI CHEBI:15603
Formula Weight 131.17
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Leucine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberB0154401 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number125125000,125121000,125120025
Lot NumberB0150359 Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number125125000,125121000,125120025
Lot NumberB0150483 Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number125125000,125121000,125120025
Lot NumberB0149340 Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number125125000,125121000,125120025
Lot NumberB0147174 Certificate TypeCertificate of Analysis Date20/10/2023 Catalog Number125125000,125121000,125120025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Leucine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.