missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Threonine, Cell Culture Reagent Product Code.: 15495889

Thermo Scientific Chemicals L-Threonine, Cell Culture Reagent

Product Code. 15495889
250 g
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy

Product Code. 15495889

Brand: Thermo Scientific Chemicals J63709.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Threonine is used to treat various nervous system disorders including spinal spasticity, multiple sclerosis, familial spastic paraparesis, and amyotrophic lateral sclerosis (ALS, Lou Gehrig's disease). Useful with intestinal disorders and indigestion, threonine has also been used to alleviate anxiety and mild depression. . L-threonine has been used clinically with the aim of increasing glycine concentrations in the cerebral spinal fluid of patients with spasticity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water 50 mg/mL.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-19-5
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064270
InChI Key AYFVYJQAPQTCCC-UHFFFAOYNA-N
Synonym l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latinShow MoreShow Less
PubChem CID 6288
ChEBI CHEBI:16857
IUPAC Name (2S,3R)-2-amino-3-hydroxybutanoic acid
SMILES CC(O)C(N)C(O)=O

Specifications

Melting Point 256°C (decomposition)
Color White
Beilstein 1721646
Merck Index 14,9380
Solubility Information Soluble in water 50mg/mL.
Quantity 250 g
Formula Weight 119.12
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Threonine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS20K051 Certificate TypeCertificate of Analysis Date27/06/2024 Catalog NumberJ63709.A1,J63709.30
Lot NumberS25G053 Certificate TypeCertificate of Analysis Date06/03/2024 Catalog NumberJ63709.A1,J63709.30
Lot NumberU11I015 Certificate TypeCertificate of Analysis Date23/08/2022 Catalog NumberJ63709.A1,J63709.30
Lot NumberR02F006 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ63709.A1,J63709.30
Lot NumberN21F027 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ63709.A1,J63709.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Threonine, Cell Culture Reagent >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.