missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Tyrosine, +99% Product Code.: 10367070

Thermo Scientific Chemicals L-Tyrosine, +99%

Product Code. 10367070
5 g, Glass Bottle
Click to view available options
Quantity:
5 g
100 g
1 kg
2.5 kg
Packaging:
Glass Bottle
Plastic Bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10367070

Brand: Thermo Scientific Chemicals 140640050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Tyrosine, CAS # 60-18-4, is an aromatic non-essential amino acid that is considered a fundamental metabolite, involved in protein synthesis and photosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• L-Tyrosine is the levorotatory isomer of the aromatic amino acid tyrosine, synthesized in vivo from L-phenylalanine
• It is available as a crystalline, off-white powder, stable at room temperature, and soluble in hydrochloric acid

Applications

• In vivo L-tyrosine is involved in protein synthesis and is a precursor to alkaloids such as morphine, neurotransmitters, epinephrine, p-coumaric acid, thyroxine, pigment melanin and catecholamines.
• Used as a supplement, L-tyrosine has been shown to increase levels of the neurotransmitters, dopamine, adrenaline, and norepinephrine in the brain
• It is considered an essential dietary supplement for phenylketonuria patients who lack the enzyme phenylalanine hydroxylase, and hence cannot convert phenylalanine into tyrosine
• L-tyrosine plays a vital role as an electron-carrier in the process of photosynthesis
• Due to its role in the synthesis of catecholamines, L-tyrosine is also used in research to study its impact on stress

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-18-4
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00002606
InChI Key OUYCCCASQSFEME-SVGMAFHSNA-N
Synonym l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, lShow MoreShow Less
PubChem CID 6057
ChEBI CHEBI:17895
IUPAC Name (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Loss on Drying 0.5% max. (1 g, 105°C)
Packaging Glass Bottle
Linear Formula 4-(HO)C6H4CH2CH(NH2)CO2H
Beilstein 14, 605
Specific Rotation -9.8° to -12.3° (20°C, 589nm) (c=5, 1N HCl)
Merck Index 15, 10020
Sulfated Ash 0.1% max.
Specific Rotation Condition − 11.05 (20.00°C c=5,dilHCL/H2O 50/50)
Quantity 5 g
Formula Weight 181.19
Percent Purity ≥99.0% (on dried substance)
Physical Form Crystalline Powder
Chemical Name or Material L-Tyrosine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0386882 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number140641000,140640010,140640025,140640050
Lot NumberA0433917 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog Number140640010,140641000,140640025,140640050
Lot NumberA0428187 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number140640010,140641000,140640025,140640050
Lot NumberA0450548 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number140640010,140641000,140640025,140640050
Lot NumberA0448392 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number140640010,140641000,140640025,140640050
compliance-icons
Product Identifier
  • L-Tyrosine
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Tyrosine, +99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.