missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Valine 99% Product Code.: 10497340

Thermo Scientific Chemicals L-Valine 99%

Product Code. 10497340
100 g, Plastic Bottle
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10497340

Brand: Thermo Scientific Chemicals 140811000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-18-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064220
InChI Key KZSNJWFQEVHDMF-BYPYZUCNSA-N
Synonym l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latinShow MoreShow Less
PubChem CID 6287
ChEBI CHEBI:16414
IUPAC Name (2S)-2-amino-3-methylbutanoic acid
SMILES CC(C)[C@H](N)C(O)=O

Specifications

Melting Point 315.0°C
Ammonium 200ppm max.
Color White
Chlorine (Cl) 200ppm max.
Foreign Amino Acids 0.5% max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 99%
Loss on Drying 0.2% max.
Packaging Plastic Bottle
Linear Formula (CH3)2CHCH(NH2)CO2H
Beilstein 04, 427
Specific Rotation + 28.00
Fieser 13,226
Sulfate 300ppm max.
Merck Index 15, 10095
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 85g/L (20°C). Other solubilities: practically insoluble in cold alcohol, ether and, acetone, soluble in mineral acids, insoluble in chloroform, diethylether
Density 1.2300g/mL
Specific Rotation Condition +28.00 (20.00°C c=8,6N HCl)
Quantity 100 g
Formula Weight 117.15
Percent Purity 99%
Specific Gravity 1.23
Physical Form Powder
Chemical Name or Material L-Valine, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468082 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number140810025,140810250,140815000,140811000
Lot NumberA0443831 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number140810025,140810250,140815000,140811000
Lot NumberA0460419 Certificate TypeCertificate of Analysis Date21/02/2024 Catalog Number140810025,140810250,140815000,140811000
Lot NumberA0451299 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number140810025,140810250,140815000,140811000
Lot NumberA0401519 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number140810025,140810250,140815000,140811000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Valine 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.