missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Lyph-Lock™ Flask Adapters

Flask Adapters: For connecting Lyph-Lock flasks to valve ports on drying chamber or manifold

Brand:  Labconco™ 7568000

 View more versions of this product

Product Code. 15173055

  • 70.55 € / Each
Estimated Shipment: 04-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Borosilicate glass, with 90° bend

Specifications

Specifications

Flask Adapter
19/38 to 0.5 in. port
Lyph lock flasks
0.5 lb.
Borosilicate Glass
For connecting lyph-lock flasks to valve ports on drying chamber or manifold
90° bend
0.2 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Labconco™ Lyph-Lock™ Flask Adapters > Standard Taper 19/38 to 1/2 in.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.