missing translation for 'onlineSavingsMsg'
Learn More
Nitritex™ Laboratory Coat Single Use Product Code.: 12952255
Estimated Shipment: 17-07-2025
to see stock.
Add to basket

Nitritex™ Laboratory Coat Single Use

Product Code. 12952255
This item is not returnable. View return policy

Product Code. 12952255

Brand: Nitritex™ BDLC2XL

Estimated Shipment: 17-07-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

The BioClean-D™ single use Lab Coat offers excellent comfort and protection. The product has been carefully developed, designed and constructed from antistatic CleanTough™ material which is particularly durable and low-linting. The garment features press stud fastenings, open cuffs, three pockets and a rear vent

Collared, open cuffs & rear vent ISO Class 4 cleanroom compatible one chest and 2 hip pockets, popper front closure white 2XL BioClean-D
TRUSTED_SUSTAINABILITY

Specifications

Product Type Controlled Environments Lab Coat
Size 2X-Large
Color White
Disposable Single-use
Sterility Non-sterile
Certifications/Compliance Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Directive 89/686/EEC
Packaging Type One piece per sealed inner PE bag; one inner bag per sealed outer PE bag; 30 outer bags per lined carton (30 pieces)
Length (Metric) 110 cm
Cleanroom Class Class 10 (ISO 4)
  • Nitritex Bioclean-D
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Nitritex Bioclean-D

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Nitritex™ Laboratory Coat Single Use >

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.