Learn More
Macherey-Nagel™ TLC Spray Visualization Reagents
Used for postchromatographic detection reactions in thin-layer chromatography. Macherey-Nagel™ TLC Spray Visualization Reagents are a selection of frequently used spray reagents that are suited for spraying or dipping TLC plates. Glass laboratory sprayer (mfr. no. 814101) sold separately.
Brand: Macherey-Nagel™ 814922
Description
- Broad selection of premium reagents available for detection of a wide range of substances
- Can be used for spraying or dipping of TLC plates
- Packaged in 100mL screw-top amber bottles
- Use with chemically-resistant sprayers, including Macherey-Nagel glass laboratory sprayer with rubber bulb (mfr. no. 814101, sold separately)
Specifications
4-(Dimethylamino)-benzaldehyde | |
Colorless | |
Spraying or Dipping | |
4-(Dimethylamino)-benzaldehyde in 2-Propanol | |
TLC plates | |
-90°C | |
Fusty, moldy | |
Spray Visualization Reagent | |
Solution |
7705-08-0 | |
0.785g/cm3 | |
0.1 to <1% 4-(Dimethylamino)-benzaldehyde | |
Detection of terpenes, sugars, steroids | |
2-Propanol | |
C9H11NO | |
Amber bottle with screw cap | |
100 mL |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- X1 4-(Dimethylamino)-benzaldeh., 100 mL
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Supplemental information
- MIXTURE LIST-Contains : 4-dimethylaminobenzaldehyde, 2-propanol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.