missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Lab Thermometer

A general purpose/stirring thermometer for a wide application of laboratory needs.

Brand:  Fisherbrand™ 63/098/1

 View more versions of this product

Product Code. 11551993

  • 6.85 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • With amber graduations
  • Non-roll fitting
  • Reinforced bulb for stirring
Specifications

Specifications

General use
305 mm
Liquid Filled Thermometer
-10°C
0.5°C
Partial Immersion
Red Spirit
50°C
-10°C to +50°C
76 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Lab Thermometer > Temperature Range: -10°C to +50°C, Subdivisions: 0.5°C, Partial Immersion, Length: 305 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.