missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ CORE Mechanical Lysis Module

The Applied Biosystems™ MagMAX™ CORE sample preparation system is a universal system designed to meet current and future veterinary testing needs.

Brand:  Applied Biosystems™ A32836

192.40 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 30057799

  • 296.00 € / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The MagMAX™ CORE Mechanical Lysis Module is offered as a supplement to the MagMAX™ CORE Nucleic Acid Purification Kit to aid in the processing of hard-to-lyse organisms like Mycobacterium avium paratuberculosis (MAP) and bovine tuberculosis in feces, milk, and environmental samples.

For a kit that contains this Mechanical Lysis Module as well as the MagMAX CORE Nucleic Acid Purification Kit, please see Cat. No. A37487.

Features include:

  • Convenience—a complete solution using the same CORE chemistry
  • Confidence—higher-quality nucleic acids from difficult-to-lyse samples, including both small (0.3 g) and large (2 to 5 g) volume feces samples
  • Flexible—purchase only what you need to lyse MAP and other hard-to-lyse organisms

The MagMAX™ CORE kit and the MagMAX™ Core Mechanical Lysis Module help make your lab more efficient through greater convenience, consistent performance, and time and cost savings.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• MagMAX CORE Clarifying Solution, 45 mL, store at room temperature
• MagMAX CORE Bead Beating Tubes, 100 tubes (2 mL), store at room temperature

Real-Time PCR, Sequencing
MagMAX™
Room Temperature
DNA (Genomic), DNA/RNA (Pathogen), Total Nucleic Acids, DNA/RNA (Viral)
High-throughput Compatible
100 tests
Bacteria, Blood, DNA, Liquid Samples (e.g. Serum), Other Samples, RNA, Tissue, Viral Samples
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2402052 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberA32836
Lot Number2402051 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberA32836
Lot Number2404053 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberA32836
Lot Number2407054 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberA32836
Lot Number2312050 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog NumberA32836
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ MagMAX™ CORE Mechanical Lysis Module > 100 tests

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.